2-Amino-6-fluorobenzaldehyde 95%

Supplier: Apollo Scientific

PC911162-1G PC911162-250MG
APOSPC911162-1GEA 133 GBP
APOSPC911162-1G APOSPC911162-250MG
2-Amino-6-fluorobenzaldehyde 95%
2-Amino-6-fluorobenzaldehyde
Formula: C₇H₆FNO
MW: 139.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12400881
CAS Number: 151585-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR