2-Chloro-3-fluoro-5-nitropyridine 98%

Supplier: Apollo Scientific

PC910735-1G PC910735-250MG PC910735-5G
APOSPC910735-1GEA 101 GBP
APOSPC910735-1G APOSPC910735-250MG APOSPC910735-5G
2-Chloro-3-fluoro-5-nitropyridine 98%
2-Chloro-3-fluoro-5-nitropyridine
Formula: C₅H₂ClFN₂O₂
MW: 176,5300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659664
CAS Number: 1079179-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR