5-Fluororesorcinol 95%

Supplier: Apollo Scientific

Synonyms: 5-Fluorobenzene-1,3-diol

PC910226-1G PC910226-500MG
APOSPC910226-1GEA 434 GBP
APOSPC910226-1G APOSPC910226-500MG
5-Fluororesorcinol 95%
5-Fluororesorcinol
Formula: C₆H₅FO₂
MW: 128,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457182
CAS Number: 75996-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR