4-(Trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carbonitrile 95%
Supplier: Apollo Scientific
PC908668-250MG
PC908668-500MG
APOSPC908668-250MGEA
374
GBP
APOSPC908668-250MG
APOSPC908668-500MG
4-(Trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carbonitrile 95%
4-(Trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carbonitrile
Formula:
C₇H₇F₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD06411653 CAS Number: 845866-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...