(2-Fluoropyridin-4-yl)methanol 98%

Supplier: Apollo Scientific

Synonyms: (2-fluoro-pyridin-4-yl)-methanol, 2-Fluoro-4-hydroxymethylpyridine

PC905392-1G PC905392-5G PC905392-10G
APOSPC905392-1GEA 31.6 GBP
APOSPC905392-1G APOSPC905392-5G APOSPC905392-10G
(2-Fluoropyridin-4-yl)methanol 98%
(2-Fluoropyridin-4-yl)methanol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200782
CAS Number: 131747-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR