[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetic acid 95%

Supplier: Apollo Scientific

PC905016-1G PC905016-250MG
APOSPC905016-1GEA 649 GBP
APOSPC905016-1G APOSPC905016-250MG
[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetic acid 95%
[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetic acid
Formula: C₇H₈F₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03419677
CAS Number: 957487-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR