(6-Fluoro-1H-indol-1-yl)acetic acid 95%

Supplier: Apollo Scientific

PC904670-1G PC904670-250MG
APOSPC904670-1GEA 571 GBP
APOSPC904670-1G APOSPC904670-250MG
(6-Fluoro-1H-indol-1-yl)acetic acid 95%
(6-Fluoro-1H-indol-1-yl)acetic acid
Formula: C₁₀H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD07401756
CAS Number: 887685-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR