5-Fluorobenzoxazole-2-thiol 98%

Supplier: Apollo Scientific

Synonyms: 5-Fluorobenzo[d]oxazole-2-thiol

PC904486-1G PC904486-5G
APOSPC904486-1GEA 119 GBP
APOSPC904486-1G APOSPC904486-5G
5-Fluorobenzoxazole-2-thiol 98%
5-Fluorobenzoxazole-2-thiol
Formula: C₇H₄FNOS
MW: 169,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08447021
CAS Number: 13451-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR