1-Chloro-2-fluoro-4-iodobenzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloro-3-fluoroiodobenzene

PC9033-100G PC9033-25G PC9033-5G
APOSPC9033-100GEA 237 GBP
APOSPC9033-100G APOSPC9033-25G APOSPC9033-5G
1-Chloro-2-fluoro-4-iodobenzene 98%
1-Chloro-2-fluoro-4-iodobenzene
Formula: C₆H₃ClFI
MW: 256,4400 g/mol
Boiling Pt: 112 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042580
CAS Number: 202982-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR