2-Bromo-6-fluoro-4-methylpyridine 98%

Supplier: Apollo Scientific

Synonyms: 2-Bromo-6-fluoro-4-picoline

PC902925-1G PC902925-5G
APOSPC902925-1GEA 77.4 GBP
APOSPC902925-1G APOSPC902925-5G
2-Bromo-6-fluoro-4-methylpyridine 98%
2-Bromo-6-fluoro-4-methylpyridine
Formula: C₆H₅BrFN
MW: 190,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839269
CAS Number: 180608-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR