5-Amino-2-fluoro-3-methylpyridine 98%

Supplier: Apollo Scientific

Synonyms: 5-Amino-2-fluoro-3-picoline, 6-Fluoro-5-methylpyridin-3-amine

PC902668-1G PC902668-25G PC902668-10G
APOSPC902668-1GEA 106 GBP
APOSPC902668-1G APOSPC902668-25G APOSPC902668-10G
5-Amino-2-fluoro-3-methylpyridine 98%
5-Amino-2-fluoro-3-methylpyridine
Formula: C₆H₇FN₂
MW: 126,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095088
CAS Number: 186593-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR