2-Fluoro-4-ethoxyphenylboronic acid 95%

Supplier: Apollo Scientific

Synonyms: (4-Ethoxy-2-fluorophenyl)boronic acid, 4-Ethoxy-2-fluorophenylboronic acid

PC901642-1G PC901642-250MG
APOSPC901642-1GEA 59.1 GBP
APOSPC901642-1G APOSPC901642-250MG
2-Fluoro-4-ethoxyphenylboronic acid 95%
2-Fluoro-4-ethoxyphenylboronic acid
Formula: C₈H₁₀BFO₃
MW: 183,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701765
CAS Number: 279261-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR