5-(4-Fluorophenyl)-1-methyl-1H-imidazol-2-ylamine 95%

Supplier: Apollo Scientific

PC901290-1G PC901290-250MG
APOSPC901290-1GEA 780 GBP
APOSPC901290-1G APOSPC901290-250MG
5-(4-Fluorophenyl)-1-methyl-1H-imidazol-2-ylamine 95%
5-(4-Fluorophenyl)-1-methyl-1H-imidazol-2-ylamine
Formula: C₁₀H₁₀FN₃
MW: 191,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457340
CAS Number: 787586-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR