[5-(Difluoromethyl)-3-methyl-1H-pyrazol-1-yl]acetic acid 95%
Supplier: Apollo Scientific
PC900732-1G
PC900732-250MG
APOSPC900732-1GEA
780
GBP
APOSPC900732-1G
APOSPC900732-250MG
[5-(Difluoromethyl)-3-methyl-1H-pyrazol-1-yl]acetic acid 95%
[5-(Difluoromethyl)-3-methyl-1H-pyrazol-1-yl]acetic acid
Formula:
C₇H₈F₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03074503 CAS Number: 512809-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...