1-(Trifluoroacetyl)piperidin-4-one 95%

Supplier: Apollo Scientific

PC900306-1G PC900306-250MG
APOSPC900306-1GEA 780 GBP
APOSPC900306-1G APOSPC900306-250MG
1-(Trifluoroacetyl)piperidin-4-one 95%
1-(Trifluoroacetyl)piperidin-4-one
Formula: C₇H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09971275
CAS Number: 65220-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR