4-Fluoroindolin-2-one 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Fluoro-2-oxindole, 4-Fluorooxindole

PC8677-1G PC8677-5G PC8677-10G
APOSPC8677-1GEA 97.1 GBP
APOSPC8677-1G APOSPC8677-5G APOSPC8677-10G
4-Fluoroindolin-2-one 97%
4-Fluorooxindole
Formula: C₈H₆FNO
MW: 151,140 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112528
CAS Number: 138343-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR