6-(2-Fluorophenyl)[1,3,5]triazine-2,4-diamine

Supplier: Apollo Scientific
Warning

PC8631-1G PC8631-25G PC8631-5G
APOSPC8631-1GEA 23.7 GBP
APOSPC8631-1G APOSPC8631-25G APOSPC8631-5G
6-(2-Fluorophenyl)[1,3,5]triazine-2,4-diamine
6-(2-Fluorophenyl)[1,3,5]triazine-2,4-diamine
Formula: C₉H₈FN₅
MW: 205,2 g/mol
Melting Pt: 243…247 °C
Storage Temperature: Ambient
MDL Number: MFCD04973088
CAS Number: 30530-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR