2-Amino-6-(trifluoromethoxy)benzoic acid

Supplier: Apollo Scientific
Warning

PC8374-1G PC8374-250MG
APOSPC8374-1GEA 525 GBP
APOSPC8374-1G APOSPC8374-250MG
2-Amino-6-(trifluoromethoxy)benzoic acid
2-Amino-6-(trifluoromethoxy)benzoic acid
Formula: C₈H₆F₃NO₃
MW: 221,13 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669643
CAS Number: 561304-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR