2-Fluoro-D-phenylalanine

Supplier: Apollo Scientific

Synonyms: (R)-2-Amino-3-(2-fluorophenyl)propanoic acid, H-D-Phe(2-F)-OH

PC8358-10G PC8358-1G PC8358-5G
APOSPC8358-10GEA 160 GBP
APOSPC8358-10G APOSPC8358-1G APOSPC8358-5G
2-Fluoro-D-phenylalanine
2-Fluoro-D-phenylalanine
Formula: C₉H₁₀FNO₂
MW: 183,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066447
CAS Number: 97731-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR