α-Bromo-4-chloro-2,6-difluorotoluene 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-Chloro-2,6-difluorobenzyl bromide, 2-(Bromomethyl)-5-chloro-1,3-difluorobenzene
PC8242-10G
PC8242-5G
PC8242-1G
PC8242-25G
APOSPC8242-10GEA
255
GBP
APOSPC8242-10G
APOSPC8242-5G
APOSPC8242-1G
APOSPC8242-25G
α-Bromo-4-chloro-2,6-difluorotoluene 98%
α-Bromo-4-chloro-2,6-difluorotoluene
Formula:
C₇H₄BrClF₂ MW: 241,46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094463 CAS Number: 537013-52-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...