2-Fluorobenzaldehyde oxime
Supplier: Apollo Scientific
PC8192-1G
PC8192-5G
APOSPC8192-1GEA
91.8
GBP
APOSPC8192-1G
APOSPC8192-5G
2-Fluorobenzaldehyde oxime
2-Fluorobenzaldehyde oxime
Formula:
C₇H₆FNO MW: 139,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00019958 CAS Number: 451-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...