2-Chloro-6-(4-fluorophenyl)nicotinonitrile 97%

Supplier: Apollo Scientific
Warning

PC8144-1G PC8144-250MG
APOSPC8144-1GEA 68.3 GBP
APOSPC8144-1G APOSPC8144-250MG
2-Chloro-6-(4-fluorophenyl)nicotinonitrile 97%
2-Chloro-6-(4-fluorophenyl)nicotinonitrile
Formula: C₁₂H₆ClFN₂
Melting Pt: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00215072
CAS Number: 31776-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR