6-Fluoroindole-3-acetonitrile

Supplier: Apollo Scientific
Warning

PC7724-100G PC7724-25G PC7724-5G
APOSPC7724-100GEA 593 GBP
APOSPC7724-100G APOSPC7724-25G APOSPC7724-5G
6-Fluoroindole-3-acetonitrile
6-Fluoroindole-3-acetonitrile
Formula: C₁₀H₇FN₂
MW: 174.18 g/mol
Melting Pt: 50…53 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09756481
CAS Number: 2341-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR