4-(4-Fluorobenzoyl)butyric acid

Supplier: Apollo Scientific
Warning

Synonyms: 5-(4-Fluorophenyl)-5-oxovaleric Acid, 5-(4-Fluorophenyl)-5-oxopentanoic acid

PC7656-100G PC7656-25G PC7656-500G
APOSPC7656-100GEA 66.9 GBP
APOSPC7656-100G APOSPC7656-25G APOSPC7656-500G
4-(4-Fluorobenzoyl)butyric acid
4-(4-Fluorobenzoyl)butyric acid
Formula: C₁₁H₁₁FO₃
MW: 210,20 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00667222
CAS Number: 149437-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR