α,α,α-Trifluoro-p-toluic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Trifluoromethyl)benzoic acid

PC7540-100G PC7540-25G PC7540-500G
APOSPC7540-100GEA 22.3 GBP
APOSPC7540-100G APOSPC7540-25G APOSPC7540-500G
α,α,α-Trifluoro-p-toluic acid 98%
α,α,α-Trifluoro-p-toluic acid
Formula: C₈H₅F₃O₂
MW: 190,12 g/mol
Boiling Pt: 247 °C/753mmHg
Melting Pt: 219…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00002562
CAS Number: 455-24-3
EINECS: 207-242-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR