4-(Trifluoromethoxy)thiobenzamide
Supplier: Apollo Scientific
|
Warning
|
PC7439U-10G
PC7439U-5G
APOSPC7439U-10GEA
360
GBP
APOSPC7439U-10G
APOSPC7439U-5G
4-(Trifluoromethoxy)thiobenzamide
4-(Trifluoromethoxy)thiobenzamide
Formula:
C₈H₆F₃NOS MW: 221.19 g/mol Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068113 CAS Number: 149169-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...