1-Chloro-4-(trifluoromethoxy)benzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Trifluoromethoxy)chlorobenzene

PC7439A-100G PC7439A-25G PC7439A-500G
APOSPC7439A-100GEA 34.1 GBP
APOSPC7439A-100G APOSPC7439A-25G APOSPC7439A-500G
1-Chloro-4-(trifluoromethoxy)benzene 98%
1-Chloro-4-(trifluoromethoxy)benzene
Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
Boiling Pt: 142…143 °C
Density: 1.367 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00040953
CAS Number: 461-81-4
EINECS: 207-315-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR