4,4,4-Trifluorobutyric acid 98%

Supplier: Apollo Scientific
Danger

PC7292-1G PC7292-25G PC7292-5G
APOSPC7292-1GEA 13.2 GBP
APOSPC7292-1G APOSPC7292-25G APOSPC7292-5G
4,4,4-Trifluorobutyric acid 98%
4,4,4-Trifluorobutyric acid
Formula: C₄H₅F₃O₂
MW: 142,08 g/mol
Boiling Pt: 166…167 °C
Melting Pt: 25…32 °C
Flash Pt: 96,1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077604
CAS Number: 406-93-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR