4-Ethoxy-2,3-difluorobenzaldehyde

Supplier: Apollo Scientific
Warning

PC7167-1G PC7167-5G PC7167-25G
APOSPC7167-1GEA 59.1 GBP
APOSPC7167-1G APOSPC7167-5G APOSPC7167-25G
4-Ethoxy-2,3-difluorobenzaldehyde
4-Ethoxy-2,3-difluorobenzaldehyde
Formula: C₉H₈F₂O₂
MW: 186,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258700
CAS Number: 126162-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR