4-Bromo-2-fluorothiobenzamide
Supplier: Apollo Scientific
PC7164-1G
PC7164-250MG
APOSPC7164-1GEA
635
GBP
APOSPC7164-1G
APOSPC7164-250MG
4-Bromo-2-fluorothiobenzamide
4-Bromo-2-fluorothiobenzamide
Formula:
C₇H₅BrFNS Storage Temperature: Ambient |
MDL Number:
MFCD08460505 CAS Number: 883230-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...