2'-Fluoro-4'-iodoacetanilide

Supplier: Apollo Scientific
Warning

PC7138-5G PC7138-1G PC7138-25G
APOSPC7138-5GEA 47.1 GBP
APOSPC7138-5G APOSPC7138-1G APOSPC7138-25G
2'-Fluoro-4'-iodoacetanilide
2'-Fluoro-4'-iodoacetanilide
Formula: C₈H₇FINO
Storage Temperature: Ambient
MDL Number: MFCD04039298
CAS Number: 97760-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR