3-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(3-Fluorophenylcarbamoyl)benzeneboronic acid

PC7083-1G PC7083-25G PC7083-5G
APOSPC7083-1GEA 68.5 GBP
APOSPC7083-1G APOSPC7083-25G APOSPC7083-5G
3-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
3-(3-Fluorophenylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₁BFNO₃
MW: 259.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027207
CAS Number: 874288-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR