2-Fluoro-5-(trifluoromethoxy)phenylboronic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Fluoro-5-(trifluoromethoxy)phenyl)boronic acid
PC6988-1G
PC6988-25G
PC6988-5G
APOSPC6988-1GEA
26.2
GBP
APOSPC6988-1G
APOSPC6988-25G
APOSPC6988-5G
2-Fluoro-5-(trifluoromethoxy)phenylboronic acid 97%
2-Fluoro-5-(trifluoromethoxy)phenylboronic acid
Formula:
C₇H₅BF₄O₃ MW: 223.9200 g/mol Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027234 CAS Number: 881402-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...