4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pentan-1-ol 95%

Supplier: Apollo Scientific
Warning

PC6932G-1G PC6932G-5G PC6932G-25G
APOSPC6932G-1GEA 95.7 GBP
APOSPC6932G-1G APOSPC6932G-5G APOSPC6932G-25G
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pentan-1-ol 95%
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pentan-1-ol
Formula: C₆H₇F₇O
Storage Temperature: Ambient
MDL Number: MFCD00155992
CAS Number: 29819-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR