2-Bromo-3-fluorobenzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC6871-25G PC6871-5G
APOSPC6871-25GEA 103 GBP
APOSPC6871-25G APOSPC6871-5G
2-Bromo-3-fluorobenzaldehyde 98%
2-Bromo-3-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782044
CAS Number: 891180-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR