Potassium fluoride, anhydrous 99%
Supplier: Apollo Scientific
|
Danger
|
PC6267S-100G
PC6267S-250G
PC6267S-500G
APOSPC6267S-100GEA
83.8
GBP
APOSPC6267S-100G
APOSPC6267S-250G
APOSPC6267S-500G
Potassium fluoride, anhydrous 99%
Potassium fluoride
Formula:
KF MW: 58,1 g/mol Boiling Pt: 1505 °C Melting Pt: 858 °C Density: 2,48 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...