(±)-1-Phenyl-2,2,2-trifluoroethanol 99%

Supplier: Apollo Scientific
Warning

Synonyms: α-(Trifluoromethyl)benzyl alcohol, 2,2,2-Trifluoro-1-phenylethan-1-ol

PC6224-1G PC6224-5G
APOSPC6224-1GEA 25 GBP
APOSPC6224-1G APOSPC6224-5G
(±)-1-Phenyl-2,2,2-trifluoroethanol 99%
(±)-1-Phenyl-2,2,2-trifluoroethanol
Formula: C₆H₅CH(OH)CF₃
MW: 176.14 g/mol
Boiling Pt: 57…59 °C/2mmHg
Density: 1.297 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00004498
CAS Number: 340-05-6
EINECS: 206-429-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR