Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC6219E-1G PC6219E-5G
APOSPC6219E-1GEA 112 GBP
APOSPC6219E-1G APOSPC6219E-5G
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride 95%
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride

Balance other HFPO oligomer acid fluorides

Formula: C₁₅F₃₀O₅
Boiling Pt: 200…203 °C
Density: 1,8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054660
CAS Number: 34761-47-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR