Ethyl-3,3,3-trifluoropropionate 97%

Supplier: Apollo Scientific
Danger

Synonyms: Ethyl 3,3,3-trifluoropropanoate

PC5769-5G PC5769-25G
APOSPC5769-5GEA 145 GBP
APOSPC5769-5G APOSPC5769-25G
Ethyl-3,3,3-trifluoropropionate 97%
Ethyl-3,3,3-trifluoropropionate
Formula: C₅H₇F₃O₂
MW: 156,11 g/mol
Boiling Pt: 78…80 °C/200mmHg
Density: 1,192 g/cm³ (25 °C)
Flash Pt: 8,5 °C
Storage Temperature: Ambient
MDL Number: MFCD01348113
CAS Number: 352-23-8
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR