4-Fluorosalicylic acid 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Fluoro-2-hydroxybenzoic acid
PC5723-1G
PC5723-5G
PC5723-10G
APOSPC5723-1GEA
13.2
GBP
APOSPC5723-1G
APOSPC5723-5G
APOSPC5723-10G
4-Fluorosalicylic acid 99%
4-Fluorosalicylic acid
Formula:
C₇H₅FO₃ MW: 156,11 g/mol Melting Pt: 186…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153168 CAS Number: 345-29-9 EINECS: 206-459-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...