4-Amino-3,5-difluorobenzonitrile

Supplier: Apollo Scientific
Warning

PC5545-1G PC5545-5G PC5545-25G
APOSPC5545-1GEA 78.7 GBP
APOSPC5545-1G APOSPC5545-5G APOSPC5545-25G
4-Amino-3,5-difluorobenzonitrile
4-Amino-3,5-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154,1169 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD03407467
CAS Number: 110301-23-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR