Potassium cyclopentyltrifluoroborate 96%
Supplier: Apollo Scientific
|
Danger
|
PC53516-25G
PC53516-5G
PC53516-1G
APOSPC53516-25GEA
107
GBP
APOSPC53516-25G
APOSPC53516-5G
APOSPC53516-1G
Potassium cyclopentyltrifluoroborate 96%
Potassium cyclopentyltrifluoroborate
Formula:
C₅H₉BF₃K Storage Temperature: Refrigerator |
MDL Number:
MFCD04112712 CAS Number: 1040745-70-7 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...