5-Fluoro-2-hydroxybenzonitrile

Supplier: Apollo Scientific
Warning

PC53108-1G PC53108-5G
APOSPC53108-1GEA 87.8 GBP
APOSPC53108-1G APOSPC53108-5G
5-Fluoro-2-hydroxybenzonitrile
5-Fluoro-2-hydroxybenzonitrile
Formula: C₇H₄FNO
MW: 137,1100 g/mol
Melting Pt: 121…126 °C
Storage Temperature: Ambient
MDL Number: MFCD11846513
CAS Number: 91407-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR