Tris[(trifluoroethoxy)methane] 97%

Supplier: Apollo Scientific
Warning

Synonyms: Tris(2,2,2-trifluoroethyl)orthoformate

PC5262-5G PC5262-1G
APOSPC5262-5GEA 362 GBP
APOSPC5262-5G APOSPC5262-1G
Tris[(trifluoroethoxy)methane] 97%
Tris[(trifluoroethoxy)methane]
Formula: C₇H₇F₉O₃
MW: 310,12 g/mol
Boiling Pt: 144…146 °C
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD04973415
CAS Number: 58244-27-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR