Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate

Supplier: Apollo Scientific

Synonyms: Ethyl 3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylate, 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylic acid Ethyl ester

PC52201-1G PC52201-5G
APOSPC52201-1GEA 98.3 GBP
APOSPC52201-1G APOSPC52201-5G
Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate
Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate
Formula: C₈H₁₀F₂N₂O₂
MW: 204,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12022341
CAS Number: 141573-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR