1-Chloro-2-fluoro-3-(2,2,2-trifluoroethyl)benzene 97%
Supplier: Apollo Scientific
|
Warning
|
PC520598-10G
PC520598-1G
PC520598-5G
APOSPC520598-10GEA
1040
GBP
APOSPC520598-10G
APOSPC520598-1G
APOSPC520598-5G
1-Chloro-2-fluoro-3-(2,2,2-trifluoroethyl)benzene 97%
1-Chloro-2-fluoro-3-(2,2,2-trifluoroethyl)benzene
Formula:
C₈H₅ClF₄ Storage Temperature: Ambient |
MDL Number:
MFCD11226522 CAS Number: 1099598-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...