(6-Fluoro-3-pyridinyl)methanol 96%

Supplier: Apollo Scientific
Warning

Synonyms: (6-Fluoropyridin-3-yl)methanol

PC520522-1G PC520522-5G
APOSPC520522-1GEA 90.5 GBP
APOSPC520522-1G APOSPC520522-5G
(6-Fluoro-3-pyridinyl)methanol 96%
(6-Fluoro-3-pyridinyl)methanol
Formula: C₆H₆FNO
MW: 127,1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095186
CAS Number: 39891-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR