5-Methyl-2-(trifluoromethyl)furo-3-ylacetonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
PC5132-1G
PC5132-250MG
APOSPC5132-1GEA
106
GBP
APOSPC5132-1G
APOSPC5132-250MG
5-Methyl-2-(trifluoromethyl)furo-3-ylacetonitrile 97%
5-Methyl-2-(trifluoromethyl)furo-3-ylacetonitrile
Formula:
C₉H₆F₃NO₂ MW: 217,15 g/mol Melting Pt: 75…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277690 CAS Number: 175276-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...