3'-(Pentafluorothio)acetophenone 97%
Supplier: Apollo Scientific
|
Warning
|
PC51188-1G
PC51188-5G
APOSPC51188-1GEA
388
GBP
APOSPC51188-1G
APOSPC51188-5G
3'-(Pentafluorothio)acetophenone 97%
3'-(Pentafluorothio)acetophenone
Formula:
C₈H₇F₅OS MW: 246,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652407 CAS Number: 159896-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...