2-Chloro-3-fluoro-4-(difluoromethyl)pyridine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-4-(difluoromethyl)-3-fluoropyridine
PC51086-1G
PC51086-250MG
PC51086-100G
PC51086-250G
APOSPC51086-1GEA
0
GBP
APOSPC51086-1G
APOSPC51086-250MG
APOSPC51086-100G
APOSPC51086-250G
2-Chloro-3-fluoro-4-(difluoromethyl)pyridine 97%
2-Chloro-3-fluoro-4-(difluoromethyl)pyridine
Formula:
C₆H₃ClF₃N MW: 181.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18384831 CAS Number: 1262413-59-1 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...